- Synthesis of vinca alkaloids and related compounds. Part 84. Sulfonamide derivatives of some vinca alkaloids with cardiovascular activity.
Synthesis of vinca alkaloids and related compounds. Part 84. Sulfonamide derivatives of some vinca alkaloids with cardiovascular activity.
Archiv der Pharmazie (1997-06-01)
I Moldvai, E Temesvári-Major, C Szántay, G Tóth, E Kárpáti, C Szántay
PMID9264244
ANOTACE
(+)-Vincamine (1) and (+)-vinpocetine (2) were chlorosulfonylated and the resulting sulfonyl chloride isomers (3-6) were transformed into sulfonamides (7-10). The ester group of sulfonamides was modified by selective hydrolysis and transesterification. Apovincaminol derivatives (14-16) were also prepared by reduction. In addition to the known cerebrovascular effects of the unsubstituted compounds (1,2) sulfonamides also show a significant peripheral vasodilator effect.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Vinpocetine impurity B, European Pharmacopoeia (EP) Reference Standard