Přejít k obsahu
Merck
  • Inhibition of UDP-GlcNAc:Gal beta 1-3GalNAc-R (GlcNAc to GalNAc) beta 6-N-acetylglucosaminyltransferase from acute myeloid leukaemia cells by photoreactive nitrophenyl substrate derivatives.
Inhibition of UDP-GlcNAc:Gal beta 1-3GalNAc-R (GlcNAc to GalNAc) beta 6-N-acetylglucosaminyltransferase from acute myeloid leukaemia cells by photoreactive nitrophenyl substrate derivatives.
Biochemical and biophysical research communications (1994-01-28)
D Toki, M A Granovsky, F Reck, W Kuhns, M A Baker, K L Matta, I Brockhausen
ANOTACE
Cells from patients with acute myeloid leukaemia (AML) contain an abnormally high UDP-GlcNAc: Gal beta 1-3GalNAc-R (GlcNAc to GalNAc) beta 6-N-acetylglucosaminyltransferase (core 2 beta 6-Gn-T) activity. Upon UV irradiation at 350 nm, the substrate Gal beta 1-3GalNAc alpha-p-nitrophenyl acted as an effective inhibitor for this enzyme but not for several other transferases. Preincubation with Gal beta 1-3GalNAc alpha-benzyl but not GalNAc alpha-benzyl protected core 2 beta 6-Gn-T from inhibition indicating that the inhibitor is specific for the substrate binding site of core 2 beta 6-Gn-T. A number of other nitrophenyl-sugar derivatives similarly acted as inhibitors for core 2 beta 6-Gn-T. GalNAc alpha-pnp at higher concentrations also inactivated UDP-Gal: GalNAc-R beta 3-galactosyltransferase from rat liver and AML cells and inhibition could be reduced by substrate protection. These results suggest that pnp-sugar derivatives may prove useful as specific inhibitors of glycosyltransferases and as affinity labels.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
p-Nitrophenyl galacto-N-bioside