Přejít k obsahu
Merck
  • Experimental studies on the carcinogenicity of five nitrogen containing polycyclic aromatic compounds directly injected into rat lungs.
Experimental studies on the carcinogenicity of five nitrogen containing polycyclic aromatic compounds directly injected into rat lungs.
Cancer letters (1983-08-01)
R P Deutsch-Wenzel, H Brune, G Grimmer
ANOTACE
Using a beeswax/tricaprylin mixture as vehicle, three doses each of acridine, benz[a]acridine (BaAC), benz[c]acridine (BcAC), dibenz[a,h]-acridine (DBa,hAC) and dibenz[a,j]acridine (DBa,jAC) were injected into the lungs of 35 female Osborne-Mendel rats per group. To compare the carcinogenic potency of the heterocycles, benzo[a]pyrene (BaP) was taken as reference substance. Dose-response relationships were obtained for DBa,hAC as well as for BaP. DBa,jAC and BcAC exhibited no carcinogenic effects in the dose range from 0.1 mg to 1 mg tested in the lung implantation model. From the results presented here, it cannot be excluded that tumor development may occur when testing higher doses. BaP, however, must be considered much more potent since it revealed, by far, higher tumor incidences and diminished life times.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Benz[a]acridine, BCR®, certified reference material