Přejít k obsahu
Merck
  • Compounds associated with algal surfaces mediate epiphytic colonization of the marine macroalga Fucus vesiculosus.
Compounds associated with algal surfaces mediate epiphytic colonization of the marine macroalga Fucus vesiculosus.
FEMS microbiology ecology (2013-01-15)
Tim Lachnit, Matthias Fischer, Sven Künzel, John F Baines, Tilmann Harder
ANOTACE
The macroalga Fucus vesiculosus carries a specific community of surface bacteria. To identify chemical compounds that possibly mediate abundance and community composition of algae-associated bacteria, we tested the effect of surface-available algal compounds on bacterial settlement and community composition under field conditions. Compounds on algal thalli were separated from the surface by extraction with organic solvents and investigated on growth inhibition and settlement of bacterial isolates. Based on in vitro data, partially purified extract fractions were then exposed to bacterial colonizers in vivo followed by bacterial enumeration and community analysis. The polar fraction of the algal surface extract revealed a significant profouling effect for Vibrionales, whereas the nonpolar fraction - containing the xanthophyll pigment fucoxanthin and other unidentified nonpolar surface compounds - revealed a significant 80% reduction of surface colonizing bacteria. The analysis of bacterial surface communities by 454 pyrosequencing demonstrated that the antifouling activity of nonpolar algal surface compounds was targeting the abundance of natural bacterial colonizers rather than the relative composition of bacterial members within the community. Moreover, the bacterial community composition on F. vesiculosus was markedly different from artificial control substrates and chemically manipulated experimental treatments, suggesting that other, nonextractable surface features and/or physical properties render algal-specific epiphytic bacterial communities.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Fucoxanthin, carotenoid antioxidant