Přejít k obsahu
Merck
Asymmetric chemoenzymatic synthesis of ramatroban using lipases and oxidoreductases.
The Journal of organic chemistry (2012-04-21)
Eduardo Busto, Vicente Gotor-Fernández, Vicente Gotor
ANOTACE
A chemoenzymatic asymmetric route for the preparation of enantiopure (R)-ramatroban has been developed for the first time. The action of lipases and oxidoreductases has been independently studied, and both were found as excellent biocatalysts for the production of adequate chiral intermediates under very mild reaction conditions. CAL-B efficiently catalyzed the resolution of (±)-2,3,4,9-tetrahydro-1H-carbazol-3-ol that was acylated with high stereocontrol. On the other hand, ADH-A mediated bioreduction of 4,9-dihydro-1H-carbazol-3(2H)-one provided an alternative access to the same enantiopure alcohol previously obtained through lipase-catalyzed resolution, a useful synthetic building block in the synthesis of ramatroban. Inversion of the absolute configuration of (S)-2,3,4,9-tetrahydro-1H-carbazol-3-ol has been identified as a key point in the synthetic route, optimizing this process to avoid racemization of the azide intermediate, finally yielding (R)-ramatroban in enantiopure form by the formation of the corresponding amine and the convenient functionalization of both exocyclic and indole nitrogen atoms.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Ramatroban, ≥98% (HPLC), solid