Přejít k obsahu
Merck
3D structure modeling of cytochrome P450 2C19 and its implication for personalized drug design.
Biochemical and biophysical research communications (2007-02-20)
Jing-Fang Wang, Dong-Qing Wei, Lin Li, Si-Yuan Zheng, Yi-Xue Li, Kuo-Chen Chou
ANOTACE
Cytochrome P450 2C19 (CYP2C19) is a member of the cytochrome P-450 enzyme superfamily and plays an important role in the metabolism of drugs. In order to gain insights for developing personalized drugs, the 3D (dimensional) structure of CYP2C19 has been developed based on the crystal structure of CYP2C9 (PDB code 1R90), and its structure-activity relationship with the ligands of CEC, Fluvoxamine, Lescol, and Ticlopidine investigated through the structure-activity relationship approach. By means of a series of docking studies, the binding pockets of CYP2C19 for the four compounds are explicitly defined that will be very useful for conducting mutagenesis studies, providing insights into personalization of drug treatments and stimulating novel strategies for finding desired personalized drugs.