Přejít k obsahu
Merck
[Biodegradability and degradation mechanism of 3-fluorophenol by the activated sludge].
Huan jing ke xue= Huanjing kexue (2006-11-30)
Chao-jie Zhang, Qi Zhou, Ling Chen, Yuan Yuan, Hui Yu
ANOTACE
An acclimated activated sludge was examined for its ability to degrade 3-fluorophenol in aerobic batch cultures. The result indicated that the organism degrades up to 100 mg/L 3-fluorophenol completely with approximately 100% fluoride anion release within 16 h. 3-Fluorophenol can serve as the sole carbon source and energy source for the organism. The acclimated activated sludge can degrade 3-fluorophenol effectively. The degradation mechanism study reveal that the initial step in the aerobic biodegradation of 3-fluorophenol is its transformation to fluorocatechol. Following transformation of the fluorophenol to fluorocatechol, ring cleavage by catechol 1, 2-dioxygenase proceedes via an ortho-cleavage pathway, then defluorination occurres.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
3-Fluorophenol, 98%