Přejít k obsahu
Merck
Biodegradation of meta-fluorophenol by an acclimated activated sludge.
Journal of hazardous materials (2006-08-16)
Zhang Chaojie, Zhou Qi, Chen Ling, Wu Zhichao, Xu Bin
ANOTACE
An acclimated activated sludge was examined for its ability to degrade meta-fluorophenol as sole carbon source in aerobic batch cultures. The mechanism study revealed that the initial step in the aerobic biodegradation of meta-fluorophenol was their transformation to fluorocatechol. Following transformation of the fluorophenol to fluorocatechol, ring cleavage by catechol 1,2-dioxygenases proceeded via an ortho-cleavage pathway, then defluorination occurred.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
3-Fluorophenol, 98%