Přejít k obsahu
Merck
  • Suppressing effects of S-methyl methanethiosulfonate and diphenyl disulfide on mitomycin C-induced somatic mutation and recombination in Drosophila melanogaster and micronuclei in mice.
Suppressing effects of S-methyl methanethiosulfonate and diphenyl disulfide on mitomycin C-induced somatic mutation and recombination in Drosophila melanogaster and micronuclei in mice.
Mutation research (1997-12-31)
Y K Nakamura, K Kawai, H Furukawa, T Matsuo, K Shimoi, I Tomita, Y Nakamura
ANOTACE
S-Methyl methanethiosulfonate (MMTS) and diphenyl disulfide (DPDS) are temporary enzyme-sulfhydryl blocking agents. They are naturally occurring phytoalexin-like and synthetic substances known to be very potent bio-antimutagens in Escherichia coli B/r WP2. In the present paper, the suppressing effects of MMTS on mitomycin C (MMC)-induced mutant wing spots in the somatic mutation and recombination test (SMART) of Drosophila melanogaster, and of MMTS and DPDS on MMC-induced micronucleated peripheral reticulocytes are described. MMTS consistently reduced the numbers of MMC-induced small single, large single and twin spots per wing at a dose of 10-1000 micrograms/vial, in a dose-dependent manner. MMTS reduced the number of twin spots per wing on the spontaneous mutation at the dose of 1000 micrograms/vial. MMTS and DPDS dose-dependently reduced the frequencies of MMC-induced micronucleated peripheral reticulocytes at a dose of 10-40, and 3-100 micrograms/kg, respectively. Our results confirmed that enzyme-sulfhydryl blocking agents, such as MMTS and DPDS, are effective antimutagens in vivo too.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Phenyl disulfide, 99%