Přejít k obsahu
Merck
  • Comparison of three rapid methods, tributyrine, 4-methylumbelliferyl butyrate, and indoxyl acetate, for rapid identification of Moraxella catarrhalis.
Comparison of three rapid methods, tributyrine, 4-methylumbelliferyl butyrate, and indoxyl acetate, for rapid identification of Moraxella catarrhalis.
Journal of clinical microbiology (1994-05-01)
E Speeleveld, J M Fossépré, B Gordts, H W Van Landuyt
ANOTACE
Moraxella catarrhalis can easily be differentiated from other oxidase-positive, gram-negative cocci with tributyrine, 4-methylumbelliferyl butyrate, or indoxyl acetate. All M. catarrhalis give positive reactions, and all Neisseria spp. give negative reactions. The 4-methylumbelliferyl butyrate tube test and indoxyl acetate strip test provide same-day identification of M. catarrhalis isolates.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Indoxyl acetate, ≥95% (GC)
Sigma-Aldrich
4-Methylumbelliferyl butyrate, suitable for fluorescence, ≥95% (HPCE)