Přejít k obsahu
Merck
  • Discrimination between UTP- and P2-purinoceptor-mediated depolarization of rat superior cervical ganglia by 4,4'-diisothiocyanatostilbene-2,2'- disulphonate (DIDS) and uniblue A.
Discrimination between UTP- and P2-purinoceptor-mediated depolarization of rat superior cervical ganglia by 4,4'-diisothiocyanatostilbene-2,2'- disulphonate (DIDS) and uniblue A.
British journal of pharmacology (1995-06-01)
G P Connolly, P J Harrison
ANOTACE
1. Using a grease-gap recording technique we have investigated the effects of some antagonists of P2-purinoceptors on the depolarization of the rat isolated superior cervical ganglion evoked by 100 microM alpha, beta-methylene-adenosine 5'-triphosphate (alpha,beta-MeATP) or uridine 5'-triphosphate (UTP). The effects of the putative P2Z-purinoceptor antagonist, coomassie brilliant blue G, putative P2X-purinoceptor antagonist, 4,4'-diisothiocyanatostilbene-2,2'-disulphonate (DIDS) and uniblue A (an analogue of the P2Y- and P2X-purinoceptor antagonist reactive blue 2) were investigated. 2. At the highest concentration examined uniblue A (300 microM) depressed alpha,beta-MeATP-induced depolarization and at 100 and 300 microM enhanced UTP-evoked depolarizations. Coomassie brilliant blue G (1 and 10 microM) did not affect depolarizations evoked by alpha,beta-MeATP or UTP. Depolarizations evoked by potassium (5 mM) or muscarine (100 nM) were unaltered by either coomassie brilliant blue G or uniblue A. Uniblue A (100 and 300 microM) produced a concentration-dependent depression of hyperpolarizations evoked by adenosine (100 microM) whereas coomassie brilliant blue G at up to 10 microM, did not alter adenosine-induced hyperpolarizations. 3. DIDS (30 and 100 microM) did not alter adenosine-evoked hyperpolarizations, or depolarizations evoked by potassium or UTP. DIDS at 100 microM did not alter depolarizations evoked by muscarine. In contrast DIDS produced a concentration-dependent depression of alpha,beta-MeATP-evoked depolarizations. 4. These results are consistent with the proposal that uniblue A and DIDS but not coomassie brilliant blue G are antagonists of P2-purinoceptors and that uniblue A is also an antagonist at P1-purinoceptors present on the rat superior cervical ganglion. 5. The ability of uniblue A and DIDS to distinguish between depolarizations evoked by UTP and alpha,beta-MeATP provides further justification for the proposal that these nucleotides activate separate receptors present on the rat superior cervical ganglion, i.e. pyrimidinoceptors and P2-purinoceptors respectively.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Uniblue A sodium salt, Reactive stain for proteins, ≥70% (HPLC)