Přejít k obsahu
Merck
  • Increased pathological complete response rate after a long-term neoadjuvant letrozole treatment in postmenopausal oestrogen and/or progesterone receptor-positive breast cancer.
Increased pathological complete response rate after a long-term neoadjuvant letrozole treatment in postmenopausal oestrogen and/or progesterone receptor-positive breast cancer.
British journal of cancer (2013-04-13)
G Allevi, C Strina, D Andreis, V Zanoni, L Bazzola, S Bonardi, C Foroni, M Milani, M R Cappelletti, F Gussago, S Aguggini, R Giardini, M Martinotti, S B Fox, A L Harris, A Bottini, A Berruti, D Generali
ANOTACE
The objective of this study was to determine the optimal scheduling of 2.5 mg daily letrozole in neoadjuvant breast cancer patients to obtain pathological complete response (pathCR) and assess Ki-67 expression as an early predictor of response. This single institution study comprised 120 oestrogen receptor (ER)-positive postmenopausal women with primary breast cancer (clinical stage ≥ T2, N0-1), from three sequential cohorts (cohort A of 40, cohort B of 40 and cohort C of 40 patients, respectively) based on different duration of the neoadjuvant letrozole. Biological markers such as ER, progesterone receptor, HER2 and Ki-67 expression were tested at diagnosis and at definitive surgery. A total of 89 patients (75.4%) achieved an objective response with 44 (37.3%) clinical CRs and 45 (38.1%) partial responses. The clinical CRs were significantly observed in cohort C (23 out of 40 patients, 57.5%) and B (16 out of 38 patients, 42.1%) compared with cohort A (5 out of 40 patients, 12.5%) (P-value for trend <0.001). Letrozole induced a similar significant reduction in Ki-67 index after treatment in all cohorts. The pathCR rate was significantly more frequent in cohort C (7 out of 40 patients, 17.5%) than in cohort A (1 out of 40 patients, 2.5%) and B (2 out of 40 patients, 5.0%) (P-value for trend <0.04). One-year neoadjuvant letrozole therapy leads to a higher pathCR rate and may be the optimal length of drug exposure.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Letrozole, ≥98% (HPLC)
Letrozole, European Pharmacopoeia (EP) Reference Standard