Přejít k obsahu
Merck
  • Postnatal switch from synaptic to extrasynaptic transmission between interneurons and NG2 cells.
Postnatal switch from synaptic to extrasynaptic transmission between interneurons and NG2 cells.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2010-05-21)
Mateo Vélez-Fort, Paloma P Maldonado, Arthur M Butt, Etienne Audinat, María Cecilia Angulo
ANOTACE
NG2 cells, oligodendrocyte precursors, play a critical role in myelination during postnatal brain maturation, but a pool of these precursors is maintained in the adult and recruited to lesions in demyelinating diseases. NG2 cells in immature animals have recently been shown to receive synaptic inputs from neurons, and these have been assumed to persist in the adult. Here, we investigated the GABAergic synaptic activity of NG2 cells in acute slices of the barrel cortex of NG2-DsRed transgenic mice during the first postnatal month, which corresponds to the period of active myelination in the neocortex. Our data demonstrated that the frequency of spontaneous and miniature GABAergic synaptic activity of cortical NG2 cells dramatically decreases after the second postnatal week, indicating a decrease in the number of synaptic inputs onto NG2 cells during development. However, NG2 cells still receive GABAergic inputs from interneurons in the adult cortex. These inputs do not rely on the presence of functional synapses but involve a form of GABA spillover. This GABA volume transmission allows interneurons to induce phasic responses in target NG2 cells through the activation of extrasynaptic GABA(A) receptors. Hence, after development is complete, volume transmission allows NG2 cells to integrate neuronal activity patterns at frequencies occurring during in vivo sensory stimulation.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
NO-711 hydrochloride, ≥98% (HPLC)