Přejít k obsahu
Merck
  • On-line preconcentration of copper as its pyrocatechol violet complex on Chromosorb 105 for flame atomic absorption spectrometric determinations.
On-line preconcentration of copper as its pyrocatechol violet complex on Chromosorb 105 for flame atomic absorption spectrometric determinations.
Journal of hazardous materials (2008-09-03)
Berrin Buke, Umit Divrikli, Mustafa Soylak, Latif Elci
ANOTACE
An on-line solid phase extraction method for the preconcentration and determination of Cu(II) by flame atomic absorption spectrometry (FAAS) has been described. It is based on the adsorption of copper(II) ion onto a home made mini column of Chromosorb 105 resin loaded with pyrocatechol violet at the pH range of 5.0-8.0, then eluted with 1 mol L(-1) HNO(3). Several parameters, such as pH of the sample solution, amount of Chromosorb 105 resin, volume of sample and eluent, type of eluent, flow rates of sample and eluent, governing the efficiency and throughput of the method were evaluated. The concentration of the copper ion detected after preconcentration was in agreement with the added amount. At optimized conditions, for 15 min of preconcentration time (30 mL of sample volume), the system achieved a detection limit of 0.02 microg L(-1), with relative standard deviation 1.1% at 0.03 microg mL(-1) copper. The present method was found to be applicable to the preconcentration of Cu(II) in natural water samples.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Pyrocatechol Violet, suitable for indicator