Přejít k obsahu
Merck
  • GABA A receptors as in vivo substrate for the anxiolytic action of valerenic acid, a major constituent of valerian root extracts.
GABA A receptors as in vivo substrate for the anxiolytic action of valerenic acid, a major constituent of valerian root extracts.
Neuropharmacology (2008-07-08)
Dietmar Benke, Andrea Barberis, Sascha Kopp, Karl-Heinz Altmann, Monika Schubiger, Kaspar E Vogt, Uwe Rudolph, Hanns Möhler
ANOTACE
Valerian extracts have been used for centuries to alleviate restlessness and anxiety albeit with unknown mechanism of action in vivo. We now describe a specific binding site on GABA(A) receptors with nM affinity for valerenic acid and valerenol, common constituents of valerian. Both agents enhanced the response to GABA at multiple types of recombinant GABA(A) receptors. A point mutation in the beta2 or beta3 subunit (N265M) of recombinant receptors strongly reduced the drug response. In vivo, valerenic acid and valerenol exerted anxiolytic activity with high potencies in the elevated plus maze and the light/dark choice test in wild type mice. In beta3 (N265M) point-mutated mice the anxiolytic activity of valerenic acid was absent. Thus, neurons expressing beta3 containing GABA(A) receptors are a major cellular substrate for the anxiolytic action of valerian extracts.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Valerenic acid, primary reference standard
Supelco
Valerenic acid, analytical standard