Přejít k obsahu
Merck
  • Correlating dynamics and selectivity in adsorption of semiconductor nanocrystals onto a self-organized pattern.
Correlating dynamics and selectivity in adsorption of semiconductor nanocrystals onto a self-organized pattern.
Nano letters (2007-10-05)
Xiaodong Chen, Michael Hirtz, Andrey L Rogach, Dmitri V Talapin, Harald Fuchs, Lifeng Chi
ANOTACE
Selective adsorption of semiconductor nanocrystals onto an organic self-organized pattern shows a time-dependent behavior. By studying the wetting behavior of delivered solvent (1-phenyloctane) on a lipid self-organized pattern and determining the adhesion energy between semiconductor nanocrystals and substrate, we obtain a correlation between dynamics and selectivity in adsorption of semiconductor nanocrystals onto the pattern by constructing a potential energy landscape. Two consecutive steps for selective adsorption of nanocrystals onto the self-organized pattern have been established: the first one is the molecule exchange of 1-phenyloctane and lipid molecules to form the adsorption sites for nanocrystals, and the second one is the adsorption of nanocrystals onto the adsorption sites due to the strong interaction between nanocrystals and substrate.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
1-Phenyloctane, 98%