Přejít k obsahu
Merck
Pharmacologic chaperones as a potential treatment for X-linked nephrogenic diabetes insipidus.
Journal of the American Society of Nephrology : JASN (2005-12-02)
Virginie Bernier, Jean-Pierre Morello, Alexandro Zarruk, Nicolas Debrand, Ali Salahpour, Michèle Lonergan, Marie-Françoise Arthus, André Laperrière, Rémi Brouard, Michel Bouvier, Daniel G Bichet
ANOTACE
In many mendelian diseases, some mutations result in the synthesis of misfolded proteins that cannot reach a transport-competent conformation. In X-linked nephrogenic diabetes insipidus, most of the mutant vasopressin 2 (V2) receptors are trapped in the endoplasmic reticulum and degraded. They are unable to reach the plasma membrane and promote water reabsorption through the principal cells of the collecting ducts. Herein is reported two types of experiments: In vivo studies to assess clinically a short-term treatment with a nonpeptide V1a receptor antagonist (SR49059) and in vitro studies in cultured cell systems. In patients, SR49059 decreased 24- h urine volume (11.9 +/- 2.3 to 8.2 +/- 2.0 L; P = 0.005) and water intake (10.7 +/- 1.9 to 7.2 +/- 1.6 L; P < 0.05). Maximum increase in urine osmolality was observed on day 3 (98 +/- 22 to 170 +/- 52 mOsm/kg; P = 0.05). Sodium, potassium, and creatinine excretions and plasma sodium were constant throughout the study. In vitro studies indicate that the nonpeptide V1a receptor antagonist SR49059 and the V1a/V2 receptor antagonist YM087 (Conivaptan) rescued cell surface expression and function of mutant V2 receptors. Mutant V2 receptors with nonsense mutations were not affected by the treatment. Misfolded V2 receptor mutants were rescued in vitro and also in vivo by nonpeptide antagonists. This therapeutic approach could be applied to the treatment of several hereditary diseases that result from errors in protein folding and kinesis.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
SR 49059, ≥97% (HPLC)