Přejít k obsahu
Merck
  • Environmental pollutant Aroclor 1242 (PCB) disrupts reproduction in adult male rhesus monkeys (Macaca mulatta).
Environmental pollutant Aroclor 1242 (PCB) disrupts reproduction in adult male rhesus monkeys (Macaca mulatta).
Environmental research (2003-11-15)
Syed Uzair Ahmad, Saeed Tariq, Samina Jalali, M Maqbool Ahmad
ANOTACE
Adult male rhesus monkeys (Macaca mulatta) were given oral treatment of either Aroclor 1242 or vehicle (corn oil and glycerol) at a dose of 200 microg/kg body wt/day for 6 months to investigate the effects of the pollutant on plasma testosterone and the morphology of testes and accessory glands. Aroclor 1242 treatment significantly decreased testicular size and testosterone levels in plasma and adversely affected spermatogenic activity by disrupting epithelial organization. All components of the germinal epithelium were greatly reduced. The spermatogonia were either hypertrophied or had shrunken vesiculated cytoplasm with distorted mitochondria and nuclear pyknosis. Changes were milder in the Sertoli cells, where nuclear infoldings were reduced. Characteristic features of treated Leydig cells were the presence of electron-dense and electron-opaque zones, appearing as plaques, cell membrane abnormalities, and high variability in nuclear shape and heterochromatin distribution. All the Aroclor 1242-treated accessory glands contained more connective tissue than their vehicle-treated counterparts. The epithelium contained many layers of irregularly shaped necrotic cells possessing stereocilia in the epididymides, either hypochromic and hypertrophied or hyperchromic and hypotrophied cells in the prostate and shrunken cuboidal cells with elongated nuclei in the seminal vesicles. In conclusion, Aroclor 1242 treatment causes severe structural alterations on gonads and accessory organs in adult male rhesus monkeys, and these effects could be mediated through both estrogen and Ah receptors.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Supelco
Aroclor 1242 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1242 solution, certified reference material, 500 mg/kg in transformer oil