Přejít k obsahu
Merck
  • Effects of heptachlor epoxide on components of various signal transduction pathways important in tumor promotion in mouse hepatoma cells. Determination of the most sensitive tumor promoter related effect induced by heptachlor epoxide.
Effects of heptachlor epoxide on components of various signal transduction pathways important in tumor promotion in mouse hepatoma cells. Determination of the most sensitive tumor promoter related effect induced by heptachlor epoxide.
Toxicology (2001-03-14)
M E Hansen, F Matsumura
ANOTACE
The effects of the organochlorine (OC) liver tumor promoter heptachlor epoxide (HE; 0, 0.1, 1, 10, and 50 microM) on several cellular tumor promoter-sensitive parameters were studied in mouse 1c1c7 hepatoma cells in an effort to identify the most sensitive biomarker for OC promoter exposure and the critical pathway and target of OC promoters. The levels of Ca2+ in the endoplasmic reticulum (ER) store, connexin43 (Cx43), PLCgamma(1), nPKCvarepsilon, and AP-1 DNA binding in nucleus were studied to screen for effects induced by submicromolar HE levels. While all the parameters tested elicited effects, particulate PLCgamma(1) and AP-1 DNA binding were found to be the most sensitive parameters affected by HE on both dose and temporal bases. Their levels were increased with 10- to 100-fold lower HE concentrations than were required to affect nPKCvarepsilon or Cx43. Further, with the lower HE dosages, particulate PLCgamma(1) and nuclear AP-1 were positively modulated by HE after 1 h versus 3 or 72 h for nPKCvarepsilon and Cx43. Ca2+ store depletion was probably the third most sensitive parameter, after AP-1 and PLCgamma(1). These results suggest the tyrosine kinase growth factor receptor pathway is the probable critical pathway for HE-induce tumor promotion with the critical target most likely being upstream of PLCgamma(1) and AP-1. This work also demonstates that upon exposure to a tumor promoter such as HE, many hepatocellular effects or changes result, suggesting that a cellular-program shift occurs similar to that described by the resistant hepatocyte model after exposure to a carcinogen or enzyme inducer.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Supelco
Heptachlor exo-epoxide, PESTANAL®, analytical standard, isomer B