Přejít k obsahu
Merck
Identification of small molecule activators of cryptochrome.
Science (New York, N.Y.) (2012-07-17)
Tsuyoshi Hirota, Jae Wook Lee, Peter C St John, Mariko Sawa, Keiko Iwaisako, Takako Noguchi, Pagkapol Y Pongsawakul, Tim Sonntag, David K Welsh, David A Brenner, Francis J Doyle, Peter G Schultz, Steve A Kay
ANOTACE
Impairment of the circadian clock has been associated with numerous disorders, including metabolic disease. Although small molecules that modulate clock function might offer therapeutic approaches to such diseases, only a few compounds have been identified that selectively target core clock proteins. From an unbiased cell-based circadian phenotypic screen, we identified KL001, a small molecule that specifically interacts with cryptochrome (CRY). KL001 prevented ubiquitin-dependent degradation of CRY, resulting in lengthening of the circadian period. In combination with mathematical modeling, our studies using KL001 revealed that CRY1 and CRY2 share a similar functional role in the period regulation. Furthermore, KL001-mediated CRY stabilization inhibited glucagon-induced gluconeogenesis in primary hepatocytes. KL001 thus provides a tool to study the regulation of CRY-dependent physiology and aid development of clock-based therapeutics of diabetes.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Longdaysin, ≥98% (HPLC)