Přejít k obsahu
Merck
  • Wogonoside inhibits lipopolysaccharide-induced angiogenesis in vitro and in vivo via toll-like receptor 4 signal transduction.
Wogonoside inhibits lipopolysaccharide-induced angiogenesis in vitro and in vivo via toll-like receptor 4 signal transduction.
Toxicology (2009-05-12)
Yan Chen, Na Lu, Yun Ling, Ying Gao, Ling Wang, Yu Sun, Qi Qi, Feng Feng, Wenyuan Liu, Wei Liu, Qidong You, Qinglong Guo
ANOTACE
Wogonoside, one flavonoid derived from the root of Scutellaria baicalensis Georgi, has been reported for its anti-inflammation activity; however, whether it can inhibit inflammation-induced angiogenesis is still unclear. In the present study, we evaluated the effect of wogonoside on lipopolysaccharide (LPS)-induced angiogenesis in vitro and in vivo. Wogonoside suppressed the LPS-stimulated migration and tube formation of human umbilical vein endothelial cells (HUVECs), as well as microvessel sprouting from rat aortic rings in vitro. Moreover, wogonoside also inhibited LPS-stimulated vessel growth of Chicken chorioallantoic membrane (CAM) in vivo. The mechanism revealed that wogonoside inhibited LPS-induced toll-like receptor 4 (TLR4) up-regulation and its downstream mitogen-activated protein kinases (MAPKs) activation, by decreasing the phosphorylation of c-Jun N-terminal kinase (JNK), extracellular signal-regulated kinase (ERK) and p38 mitogen-activated protein kinase. The results suggest that wogonoside inhibits LPS-induced angiogenesis both in vitro and in vivo, and that it might have a therapeutic potential for the diseases associated with the development of both inflammation and angiogenesis progress.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Wogonoside, ≥95% (LC/MS-ELSD)