Přejít k obsahu
Merck
  • Transcriptional activation of pig Galβ1,3GalNAc α2,3-sialyltransferase (pST3Gal I) gene by TGF-β1 in porcine kidney PK-15 cells.
Transcriptional activation of pig Galβ1,3GalNAc α2,3-sialyltransferase (pST3Gal I) gene by TGF-β1 in porcine kidney PK-15 cells.
Biochemical and biophysical research communications (2011-09-29)
Sung-Wook Son, Kwon-Ho Song, Haw-Young Kwon, Kyoung-Sook Kim, Cheorl-Ho Kim, Keon-Bong Oh, Young-Kug Choo, Young-Choon Lee
ANOTACE
In this study we investigated for the first time the transcriptional regulation of pig Galβ1,3GalNAc α2,3-sialyltransferase (pST3Gal I) in response to TGF-β1 in porcine kidney PK-15 cells. The pST3Gal I gene was found to span about 90kb and to be composed of 8 exons including 2 exons in the 5'-untranslated region. RT-PCR analysis indicated that the induction of pST3Gal I by TGF-β1 is regulated at the transcriptional level. Functional analysis of the 5'-flanking region of the pST3Gal I gene revealed the -1257 to -976 region functions as the TGF-β1-inducible promoter and that the Smad-binding site at -1020 is crucial for TGF-β1-induced expression of pST3Gal I in PK-15 cells. In addition, the transcriptional activity of pST3Gal I induced by TGF-β1 in PK-15 cells was strongly inhibited by SIS3, which is a specific Smad-3 inhibitor. In summary, our results identified the core promoter region in the pST3Gal I promoter and demonstrated that Smad-3 binding to the Samd-3 binding site at -1020 is essential for transcriptional activation of pST3Gal I in TGF-β1-induced PK-15 cells.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
α-2,3-Sialyltransferase from Pasteurella multocida, recombinant, expressed in E. coli BL21, ≥2 units/mg protein