Přejít k obsahu
Merck
  • Multiscale reorganization of the genome following DNA damage facilitates chromosome translocations via nuclear actin polymerization.
Multiscale reorganization of the genome following DNA damage facilitates chromosome translocations via nuclear actin polymerization.
Nature structural & molecular biology (2022-12-24)
Jennifer Zagelbaum, Allana Schooley, Junfei Zhao, Benjamin R Schrank, Elsa Callen, Shan Zha, Max E Gottesman, André Nussenzweig, Raul Rabadan, Job Dekker, Jean Gautier
ANOTACE
Nuclear actin-based movements have been shown to orchestrate clustering of DNA double-strand breaks (DSBs) into homology-directed repair domains. Here we describe multiscale three-dimensional genome reorganization following DNA damage and analyze the contribution of the nuclear WASP-ARP2/3-actin pathway toward chromatin topology alterations and pathologic repair. Hi-C analysis reveals genome-wide, DNA damage-induced chromatin compartment flips facilitated by ARP2/3 that enrich for open, A compartments. Damage promotes interactions between DSBs, which in turn facilitate aberrant, actin-dependent intra- and inter-chromosomal rearrangements. Our work establishes that clustering of resected DSBs into repair domains by nuclear actin assembly is coordinated with multiscale alterations in genome architecture that enable homology-directed repair while also increasing nonhomologous end-joining-dependent translocation frequency.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Wiskostatin
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer