Přejít k obsahu
Merck
  • Functional mapping of PHF6 complexes in chromatin remodeling, replication dynamics, and DNA repair.
Functional mapping of PHF6 complexes in chromatin remodeling, replication dynamics, and DNA repair.
Blood (2022-03-27)
Silvia Alvarez, Ana C da Silva Almeida, Robert Albero, Mayukh Biswas, Angelica Barreto-Galvez, Thomas S Gunning, Anam Shaikh, Tomas Aparicio, Agnieszka Wendorff, Erich Piovan, Pieter Van Vlierberghe, Steven Gygi, Jean Gautier, Advaitha Madireddy, Adolfo A Ferrando
ANOTACE
The Plant Homeodomain 6 gene (PHF6) encodes a nucleolar and chromatin-associated leukemia tumor suppressor with proposed roles in transcription regulation. However, specific molecular mechanisms controlled by PHF6 remain rudimentarily understood. Here we show that PHF6 engages multiple nucleosome remodeling protein complexes, including nucleosome remodeling and deacetylase, SWI/SNF and ISWI factors, the replication machinery and DNA repair proteins. Moreover, after DNA damage, PHF6 localizes to sites of DNA injury, and its loss impairs the resolution of DNA breaks, with consequent accumulation of single- and double-strand DNA lesions. Native chromatin immunoprecipitation sequencing analyses show that PHF6 specifically associates with difficult-to-replicate heterochromatin at satellite DNA regions enriched in histone H3 lysine 9 trimethyl marks, and single-molecule locus-specific analyses identify PHF6 as an important regulator of genomic stability at fragile sites. These results extend our understanding of the molecular mechanisms controlling hematopoietic stem cell homeostasis and leukemia transformation by placing PHF6 at the crossroads of chromatin remodeling, replicative fork dynamics, and DNA repair.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-PHF6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody