Přejít k obsahu
Merck
  • Discovery of Novel Potent Reversible and Irreversible Myeloperoxidase Inhibitors Using Virtual Screening Procedure.
Discovery of Novel Potent Reversible and Irreversible Myeloperoxidase Inhibitors Using Virtual Screening Procedure.
Journal of medicinal chemistry (2017-07-04)
Jalal Soubhye, Ibaa Chikh Alard, Iyas Aldib, Martine Prévost, Michel Gelbcke, Annelise De Carvalho, Paul G Furtmüller, Christian Obinger, Jörg Flemmig, Sara Tadrent, Franck Meyer, Alexandre Rousseau, Jean Nève, Véronique Mathieu, Karim Zouaoui Boudjeltia, François Dufrasne, Pierre Van Antwerpen
ANOTACE
The heme enzyme myeloperoxidase (MPO) participates in innate immune defense mechanism through formation of microbicidal reactive oxidants. However, evidence has emerged that MPO-derived oxidants contribute to propagation of inflammatory diseases. Because of the deleterious effects of circulating MPO, there is a great interest in the development of new efficient and specific inhibitors. Here, we have performed a novel virtual screening procedure, depending on ligand-based pharmacophore modeling followed by structure-based virtual screening. Starting from a set of 727842 compounds, 28 molecules were selected by this virtual method and tested on MPO in vitro. Twelve out of 28 compounds were found to have an IC50 less than 5 μM. The best inhibitors were 2-(7-methoxy-4-methylquinazolin-2-yl)guanidine (28) and (R)-2-(1-((2,3-dihydro-1H-imidazol-2-yl)methyl)pyrrolidin-3-yl)-5-fluoro-1H-benzo[d]imidazole (42) with IC50 values of 44 and 50 nM, respectively. Studies on the mechanism of inhibition suggest that 28 is the first potent mechanism-based inhibitor and inhibits irreversibly MPO at nanomolar concentration.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Compound 18, ≥98% (HPLC)