Přejít k obsahu
Merck
Rapid development of motion-streak coding in the mouse visual cortex.
iScience (2023-01-04)
Manavu Tohmi, Jianhua Cang
ANOTACE
Despite its importance, the development of higher visual areas (HVAs) at the cellular resolution remains largely unknown. Here, we conducted 2-photon calcium imaging of mouse HVAs lateromedial (LM) and anterolateral (AL) and V1 to observe developmental changes in visual response properties. HVA neurons showed selectivity for orientations and directions similar to V1 neurons at eye opening, which became sharper in the following weeks. Neurons in all areas over all developmental stages tended to respond selectively to dots moving along an axis perpendicular to their preferred orientation at slow speeds, suggesting a certain level of conventional motion coding already at eye opening. In contrast, at high speeds, many neurons responded to dots moving along the axis parallel to the preferred orientation in older animals but rarely after eye opening, indicating a lack of motion-streak coding in the earlier stage. Together, our results uncover the development of visual properties in HVAs.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Chlorprothixene hydrochloride