Přejít k obsahu
Merck
Generation of PCBP1-deficient pigs using CRISPR/Cas9-mediated gene editing.
iScience (2022-10-25)
Chunyun Qi, Daxin Pang, Kang Yang, Shuyu Jiao, Heyong Wu, Chuheng Zhao, Lanxin Hu, Feng Li, Jian Zhou, Lin Yang, Dongmei Lv, Xiaochun Tang, Hongsheng Ouyang, Zicong Xie
ANOTACE
Classical swine fever virus (CSFV), a classic swine fever pathogen, causes severe economic losses worldwide. Poly (rC)-binding protein 1 (PCBP1), which interacts with Npro of CSFV, plays a vital role in CSFV growth. We are the first to report the generation of PCBP1-deficient pigs via gene-editing technology. The PCBP1-deficient pigs exhibited normal birth weight and reproductive-performance traits and developed normally. Viral challenge experiments indicated that primary cells isolated from F0- and F1-generation pigs exhibited significantly reduced CSFV infection. Additional mechanistic exploration further confirmed that the PCBP1 deficiency-mediated antiviral effect is related to the activation of type I interferon (IFN). Besides showing that a gene-editing strategy could be used to generate PCBP1-deficient pigs, our study introduces a valuable animal model for further investigating the infection mechanisms of CSFV that will help to develop better antiviral solutions.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-Pig IgG (whole molecule)−FITC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution