Přejít k obsahu
Merck
  • Serum Profiling for Identification of Autoantibody Signatures in Diseases Using Protein Microarrays.
Serum Profiling for Identification of Autoantibody Signatures in Diseases Using Protein Microarrays.
Methods in molecular biology (Clifton, N.J.) (2017-07-05)
Shabarni Gupta, K P Manubhai, Shuvolina Mukherjee, Sanjeeva Srivastava
ANOTACE
Protein microarrays are platforms for studying protein-protein interactions and identifying disease-related self-antigens/autoantigens, which elicit an immune response in a high-throughput format. Protein arrays have been extensively used over the past two decades for several clinical applications. By using this platform, serum containing autoantibodies against potential self-antigens can be screened on proteome-wide arrays, harboring a large repertoire of full-length human proteins. Identification of such autoantigens can help deducing early diagnostic, as well as, prognostic markers in case of malignancies, autoimmune disorders, and other systemic diseases. Here, we provide an overview of the protein microarray technology along with details of an established method to study autoantibody profiles from patient sera.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-Glutathione-S-Transferase Antibody, S. japonicum form, Chemicon®, from rabbit