Přejít k obsahu
Merck
Evaluation of MAGE-D4 expression in hepatocellular carcinoma in Japanese patients.
Journal of surgical oncology (2013-09-27)
Hideki Takami, Mitsuro Kanda, Hisaharu Oya, Soki Hibino, Hiroyuki Sugimoto, Masaya Suenaga, Suguru Yamada, Yoko Nishikawa, Mikako Asai, Tsutomu Fujii, Shuji Nomoto, Yasuhiro Kodera
ANOTACE
Though Melanoma-associated antigen (MAGE) family genes have received lots of attention as cancer-related genes and targets for immunotherapy, MAGE-D4 expression in hepatocellular carcinoma (HCC) has not yet been evaluated. MAGE-D4 mRNA expression was assayed in nine HCC cell lines and 94 HCC surgical specimens obtained from Japanese patients by quantitative real-time reverse transcription polymerase chain reaction, and the correlations between MAGE-D4 mRNA expression and clinicopathological factors were evaluated. The expression and distribution of MAGE-D4b protein were evaluated immunohistochemically. MAGE-D4 mRNA was overexpressed in five of nine HCC cell lines and 34 of 94 primary HCCs (36.2%). Median overall survival (14.8 vs. 118 months, P < 0.001) and relapse-free survival (2.7 vs. 18.3 months, P < 0.001) were significantly shorter in patients with high than with low-moderate MAGE-D4 expression. Multivariate analysis for overall survival showed that MAGE-D4 overexpression was independently prognostic for survival (hazard ratio 2.88, P = 0.009) and significantly associated with high alpha-fetoprotein concentration (P < 0.001), poor tumor differentiation (P = 0.003) and vascular invasion (P = 0.021). MAGE-D4b protein expression patterns were consistent with those of MAGE-D4 mRNA. Overexpression of MAGE-D4 may be a predictive marker of early recurrence and mortality in patients with HCC.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-MAGED4B antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution