Přejít k obsahu
Merck
  • A vasculature niche orchestrates stromal cell phenotype through PDGF signaling: Importance in human fibrotic disease.
A vasculature niche orchestrates stromal cell phenotype through PDGF signaling: Importance in human fibrotic disease.
Proceedings of the National Academy of Sciences of the United States of America (2022-03-24)
Thomas B Layton, Lynn Williams, Nan Yang, Mingjun Zhang, Carl Lee, Marc Feldmann, Glenda Trujillo, Dominic Furniss, Jagdeep Nanchahal
ANOTACE
SignificanceTissue fibrotic diseases, for example of the liver and lung, represent a huge unmet medical need. In this study, using single-cell RNA sequencing, cytometry by time of flight (CyTOF), tissue imaging, and functional assays, we identify a complex vascular niche in Dupuytren's disease (DD), a common localized fibrotic condition of the palm, where early-disease-stage tissue can be accessed readily. We uncover a population of myofibroblast precursors within the pericyte compartment and demonstrate that the endothelium instructs the differentiation of functionally distinct stromal cells, thereby orchestrating discrete microenvironments in the fibrotic milieu. Together, these findings provide a basis for the concept of targeting blood vessel signaling to control the progression of human fibrosis.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Monoclonal Anti-CD140b antibody produced in mouse, clone 18A2, purified immunoglobulin, buffered aqueous solution