Přejít k obsahu
Merck
Vitamin D supplementation modulates autophagy in the pristane-induced lupus model.
Advances in rheumatology (London, England) (2022-07-23)
Manuela Dos Santos, Jordana Miranda de Souza Silva, Bárbara Jonson Bartikoski, Eduarda Correa Freitas, Amanda Busatto, Rafaela Cavalheiro do Espírito Santo, Odirlei Andre Monticielo, Ricardo Machado Xavier
ANOTACE
Clinical evidence of skeletal muscle involvement is not uncommon in systemic lupus erythematosus (SLE). Because of the poor understanding of signaling pathways involved in SLE muscle wasting, the  aim of this study was to evaluate the effects of vitamin D supplementation on skeletal muscle in mice with pristane-induced lupus. Balb/c mice with lupus-like disease induced by pristane injection were randomized into three groups: pristane-induced lupus (PIL; n = 10), pristane-induced lupus + vitamin D supplementation (PIL + VD; n = 10) and healthy controls (CO; n = 8). Physical function was evaluated on days 0, 60, 120 and 180. The tibialis anterior and gastrocnemius muscles were collected to evaluate myofiber cross-sectional area (CSA) and protein expression. The PIL + VD group showed lower muscle strength compared to the CO and PIL groups at different time points. PIL mice showed similar myofiber CSA compared to CO and PIL + VD groups. LC3-II expression was higher in PIL compared to CO and PIL + VD groups. MyoD expression was higher in PIL mice compared to PIL + VD, while myostatin expression was higher in PIL + VD than PIL group. Myogenin expression levels were decreased in the PIL + VD group compared with the CO group. The Akt, p62 and MuRF expressions and mobility assessment showed no significance. Changes in skeletal muscle in PIL model happen before CSA reduction, possibly due to autophagy degradation, and treatment with Vitamin D has a impact on physical function by decreasing muscle strength and time of fatigue.. Vitamin D supplementation has a potential role modulating physical parameters and signaling pathways in muscle during pristane-induced lupus model.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-MYOD1 (Ab-200) antibody produced in rabbit, affinity isolated antibody