Přejít k obsahu
Merck
  • Discovery and SAR studies of 3-amino-4-(phenylsulfonyl)tetrahydrothiophene 1,1-dioxides as non-electrophilic antioxidant response element (ARE) activators.
Discovery and SAR studies of 3-amino-4-(phenylsulfonyl)tetrahydrothiophene 1,1-dioxides as non-electrophilic antioxidant response element (ARE) activators.
Bioorganic chemistry (2021-01-29)
Jeyun Jo, Lara Ibrahim, Jonathan Iaconelli, Jinsook Kwak, Manoj Kumar, Yunjin Jung, Luke L Lairson, Arnab K Chatterjee, Peter G Schultz, Michael J Bollong, Hwayoung Yun
ANOTACE
The transcription factor NRF2 controls resistance to oxidative insult and is thus a key therapeutic target for treating a number of disease states associated with oxidative stress and aging. We previously reported CBR-470-1, a bis-sulfone which activates NRF2 by increasing the levels of methylglyoxal, a metabolite that covalently modifies NRF2 repressor KEAP1. Here, we report the design, synthesis, and structure activity relationship of a series of bis-sulfones derived from this unexplored chemical template. We identify analogs with sub-micromolar potencies, 7f and 7g, as well as establish that efficacious NRF2 activation can be achieved by non-toxic analogs 7c, 7e, and 9, a key limitation with CBR-470-1. Further efforts to identify non-covalent NRF2 activators of this kind will likely provide new insight into revealing the role of central metabolism in cellular signaling.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
CBR-470-1, racemic, ≥98% (HPLC)