Přejít k obsahu
Merck
  • Cellular Adhesion Is a Controlling Factor in Neutrophil Extracellular Trap Formation Induced by Anti-Neutrophil Cytoplasmic Antibodies.
Cellular Adhesion Is a Controlling Factor in Neutrophil Extracellular Trap Formation Induced by Anti-Neutrophil Cytoplasmic Antibodies.
ImmunoHorizons (2022-02-24)
Patrick M Lelliott, Masayuki Nishide, Nicolas Pavillon, Yasutaka Okita, Takayuki Shibahara, Yumiko Mizuno, Hanako Yoshimura, Sho Obata, Atsushi Kumanogoh, Nicholas I Smith
ANOTACE
Anti-neutrophil cytoplasmic Ab (ANCA)-associated vasculitis (AAV) is a life-threatening condition characterized by improper activation of neutrophils and the release of neutrophil extracellular traps (NETs) in small vessels. This study aimed to explain the role of NETs in AAV pathogenesis by investigating a link between adhesion and NET release using human neutrophils. We leveraged an imaging flow cytometry-based assay and three-dimensional culture to demonstrate that neutrophil adhesion is essential for ANCA-induced NET formation. We confirmed this requirement for cell adhesion using standard microscopy on ultra-low attachment hydrogel surfaces and demonstrate that this depends on the focal adhesion kinase pathway as determined using inhibitors for multiple targets in this process. ANCA increased expression of β2 integrins on neutrophils, and we confirmed that these integrins were required for NET formation using blocking Abs. Finally, inhibitors for oxidative burst prevented NET formation, and this oxidative burst was mediated by the focal adhesion pathway. Overall, our findings reveal a central role for neutrophil attachment in NET formation in response to ANCAs, helping to explain the restricted localization pattern of vessel damage, and suggesting that targeting neutrophil adhesion factors may be beneficial in preventing pathological damage from NETs during AAV.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-Integrin α9β1 Antibody, clone Y9A2, azide free, clone Y9A2, Chemicon®, from mouse