Přejít k obsahu
Merck
  • αvβ3 Integrin Expression and Mitogenic Effects by Thyroid Hormones in Chronic Lymphocytic Leukemia.
αvβ3 Integrin Expression and Mitogenic Effects by Thyroid Hormones in Chronic Lymphocytic Leukemia.
Journal of clinical medicine (2021-05-01)
Uri Abadi, Avivit Weisz, Dvora Kidron, Aviva Katzav, Aleck Hercbergs, Paul J Davis, Martin H Ellis, Osnat Ashur-Fabian
ANOTACE
Chronic lymphocytic leukemia (CLL) is the most common adult leukemia. The thyroid hormones, T3 and T4, bind the αvβ3 integrin and activate phosphorylates ERK (pERK). These tumor-promoting actions were reported in a number of malignancies, but not in CLL. Primary cells from 22 CLL patients were verified for disease markers (CD5/CD19/CD23) and analyzed for αvβ3 by flow cytometry (FC), ImageStream, Western blots (WB), and immunohistochemistry (IHC) in archival bone marrow (BM, n = 6) and lymph node (LN, n = 5) tissues. Selected samples (n = 8) were incubated with T3 (1-100 nM) or T4 (0.1-10 µM) for 30 min, and the expression levels of αvβ3, pERK and PCNA (cell proliferation marker) were determined (WB). αvβ3 was detected on the membrane of circulating CLL cells and in the BM but not in the LN. T3 and T4 enhanced αvβ3 protein levels in primary CLL cells. Similarly, pERK and PCNA were rapidly induced in response to T3 and T4 exposure. αvβ3 integrin is expressed on primary CLL cells and is induced by thyroid hormones. We further suggest that the hormones are mitogenic in these cells, presumably via αvβ3-mediated signaling.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, FITC conjugated, clone LM609, Chemicon®, from mouse