Přejít k obsahu
Merck
Nemaline myopathy caused by absence of alpha-skeletal muscle actin.
Annals of neurology (2006-12-26)
Kristen J Nowak, Caroline A Sewry, Carmen Navarro, Waney Squier, Cristina Reina, Jose R Ricoy, Sandeep S Jayawant, Anne-Marie Childs, J Angus Dobbie, Richard E Appleton, Roger C Mountford, Kendall R Walker, Sophie Clement, Annie Barois, Francesco Muntoni, Norma B Romero, Nigel G Laing
ANOTACE
To investigate seven congenital myopathy patients from six families: one French Gypsy, one Spanish Gypsy, four British Pakistanis, and one British Indian. Three patients required mechanical ventilation from birth, five died before 22 months, one is ventilator-dependent, but one, at 30 months, is sitting with minimal support. All parents were unaffected. The alpha-skeletal muscle actin gene (ACTA1) was sequenced. Available muscle biopsies were investigated by standard histological and electron microscopic techniques. The expression of various proteins was determined by immunohistochemistry, western blotting, or both. Three homozygous ACTA1 null mutations were identified: p.Arg41X in the French patient, p.Tyr364fsX in the Spanish patient, and p.Asp181fsX10 in all five British patients. An absence of alpha-skeletal muscle actin protein but presence of alpha-cardiac actin was shown in all muscle biopsies examined, with more alpha-cardiac actin in the biopsy from the child with the greatest muscle function. Muscle biopsies from all patients exhibited nemaline bodies whereas three also contained zebra bodies. The seven patients have recessive nemaline myopathy caused by absence of alpha-skeletal muscle actin. The level of retention of alpha-cardiac actin, the skeletal muscle fetal actin isoform, may determine alpha-skeletal muscle actin disease severity. This has implications for possible future therapy.