Přejít k obsahu
Merck
  • The murine orthologue of the Golgi-localized TPTE protein provides clues to the evolutionary history of the human TPTE gene family.
The murine orthologue of the Golgi-localized TPTE protein provides clues to the evolutionary history of the human TPTE gene family.
Human genetics (2002-01-26)
M Guipponi, C Tapparel, O Jousson, N Scamuffa, C Mas, C Rossier, P Hutter, P Meda, R Lyle, A Reymond, S E Antonarakis
ANOTACE
The human TPTE gene encodes a testis-specific protein that contains four potential transmembrane domains and a protein tyrosine phosphatase motif, and shows homology to the tumor suppressor PTEN/MMAC1. Chromosomal mapping revealed multiple copies of the TPTE gene present on the acrocentric chromosomes 13, 15, 21 and 22, and the Y chromosome. Zooblot analysis suggests that mice may possess only one copy of TPTE. In the present study, we report the isolation and initial characterization of the full-length cDNA of the mouse homologue Tpte. At least three different mRNA transcripts ( Tpte.a, b, c) are produced via alternative splicing, encoding predicted proteins that would contain four potential transmembrane domains and a protein tyrosine phosphatase motif. Transfection of a 5'EGFP-TPTE fusion protein in Hela cells revealed an intracellular localization within the Golgi apparatus. Tpte was mapped by radiation hybrid to a region of mouse chromosome 8 that shows conserved synteny with human 13q14.2-q21 between NEK3 and SGT1. This region of the human genome was found to contain a partial, highly diverged copy of TPTE that is likely to represent the ancestral copy from which the other copies of TPTE arose through duplication events. The Y chromosome copy of TPTE is a pseudogene and is not therefore involved in the testis expression of this gene family.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine, 96%