Přejít k obsahu
Merck
  • Inhibition of CINC-1 decreases right ventricular damage caused by experimental pulmonary embolism in rats.
Inhibition of CINC-1 decreases right ventricular damage caused by experimental pulmonary embolism in rats.
Journal of immunology (Baltimore, Md. : 1950) (2007-11-21)
John Zagorski, Michael A Gellar, Maria Obraztsova, Jeffrey A Kline, John A Watts
ANOTACE
Right ventricular (RV) dysfunction is a strong risk factor for poor clinical outcome following pulmonary embolism (PE), the third most prevalent cardiovascular disease. Previous studies in our laboratory demonstrated that RV failure during PE is mediated, in part, by neutrophil-dependant cardiac inflammation. In this study we use DNA microarray analysis of gene expression to demonstrate that PE results in increased expression of the CXC chemokines CINC-1 and CINC-2 between 6 and 18 h after the start of PE in a rat model of PE. Neutrophils accumulate in RV tissue by 18 h, and this inflammation is associated with decreased right heart function. Treatment of rats with Abs to CINC-1 significantly suppressed neutrophil accumulation in RVs during PE (52% reduction in tissue myeloperoxidase) and ameliorated RV failure. In addition, plasma concentration of cardiac troponin I, an established diagnostic marker for cardiac damage, was reduced by 90%. These results suggest that selective anti-inflammatory therapies targeted at neutrophil chemoattractants will reduce cardiac inflammation and reduce RV damage in the setting of PE.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Leukocyte Naphthol AS-D Chloroacetate (Specific Esterase) Kit