Přejít k obsahu
Merck
Hypothalamic-Extended Amygdala Circuit Regulates Temporal Discounting.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2021-01-15)
Haofang E Li, Mark A Rossi, Glenn D R Watson, H Gregory Moore, Min Tong Cai, Namsoo Kim, Katrina A Vokt, Dongye Lu, Ryan A Bartholomew, Ryan N Hughes, Henry H Yin
ANOTACE
Choice behavior is characterized by temporal discounting, i.e., preference for immediate rewards given a choice between immediate and delayed rewards. Agouti-related peptide (AgRP)-expressing neurons located in the arcuate nucleus of the hypothalamus (ARC) regulate food intake and energy homeostasis, yet whether AgRP neurons influence choice behavior and temporal discounting is unknown. Here, we demonstrate that motivational state potently modulates temporal discounting. Hungry mice (both male and female) strongly preferred immediate food rewards, yet sated mice were largely indifferent to reward delay. More importantly, selective optogenetic activation of AgRP-expressing neurons or their axon terminals within the posterior bed nucleus of stria terminalis (BNST) produced temporal discounting in sated mice. Furthermore, activation of neuropeptide Y (NPY) type 1 receptors (Y1Rs) within the BNST is sufficient to produce temporal discounting. These results demonstrate a profound influence of hypothalamic signaling on temporal discounting for food rewards and reveal a novel circuit that determine choice behavior.SIGNIFICANCE STATEMENT Temporal discounting is a universal phenomenon found in many species, yet the underlying neurocircuit mechanisms are still poorly understood. Our results revealed a novel neural pathway from agouti-related peptide (AgRP) neurons in the hypothalamus to the bed nucleus of stria terminalis (BNST) that regulates temporal discounting in decision-making.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-NPY1R antibody produced in rabbit, affinity isolated antibody