Přejít k obsahu
Merck
Leptin in immunology.
Journal of immunology (Baltimore, Md. : 1950) (2005-03-08)
Giuseppe Matarese, Stergios Moschos, Christos S Mantzoros
ANOTACE
Leptin is an adipokine which conveys information on energy availability. In humans, leptin influences energy homeostasis and regulates neuroendocrine function primarily in states of energy deficiency. As a cytokine, leptin also affects thymic homeostasis and, similar to other proinflammatory cytokines, leptin promotes Th1 cell differentiation and cytokine production. We review herein recent advances on the role of leptin in the pathophysiology of immune responses.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Millipore
Rat Leptin ELISA, This Rat Leptin ELISA is used to measure & quantify Leptin levels in Metabolism & Endocrine research.