Přejít k obsahu
Merck
  • Characterization of calcineurin in human neutrophils. Inhibitory effect of hydrogen peroxide on its enzyme activity and on NF-kappaB DNA binding.
Characterization of calcineurin in human neutrophils. Inhibitory effect of hydrogen peroxide on its enzyme activity and on NF-kappaB DNA binding.
The Journal of biological chemistry (1998-12-29)
M Carballo, G Márquez, M Conde, J Martín-Nieto, J Monteseirín, J Conde, E Pintado, F Sobrino
ANOTACE
We describe here a specific calcineurin activity in neutrophil lysates, which is dependent on Ca2+, inhibited by trifluoroperazine, and insensitive to okadaic acid. Immunoblotting experiments using a specific antiserum recognized both the A and B chains of calcineurin. Neutrophils treated with cyclosporin A or FK 506 showed a dose-dependent inhibition of calcineurin activity. The effect of oxidant compounds on calcineurin activity was also investigated. Neutrophils treated with hydrogen peroxide (H2O2), where catalase was inhibited with aminotriazole, exhibited a specific inhibition of calcineurin activity. However, the addition of reducing agents to neutrophil extracts partially reversed the inhibition caused by H2O2. A similar inhibitory effect of H2O2 on calcineurin activity was observed to occur in isolated lymphocytes. This is the first demonstration that redox agents modulate calcineurin activity in a cellular system. In addition, electrophoretic mobility shift assays revealed that lipopolysaccharide-induced activation of NF-kappaB in human neutrophils is inhibited by cell pretreatment with H2O2 in a dose-dependent manner. These data indicate that calcineurin activity regulates the functional activity of lipopolysaccharide-induced NF-kappaB/Rel proteins in human neutrophils. These data indicate a role of peroxides in the modulation of calcineurin activity and that the H2O2-dependent NF-kappaB inactivation in neutrophils occurs in concert with inhibition of calcineurin.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Trifluoperazine dihydrochloride, ≥99%, powder