Přejít k obsahu
Merck
Monitoring damage of self-assembled monolayers using metastable excited helium atoms.
The Journal of chemical physics (2021-01-28)
Georgios Stratis, Jordan D Zesch, Henry S Pan, Lauren J Webb, Mark G Raizen
ANOTACE
The breaking of molecular bonds during exposure to ionizing radiation and electron beams creates irreversible damage in the molecular structure. In some cases, such as lithography, controlled damage of a molecular resist is a desirable process and is the basis for the entire semiconductor industry. In other cases, such as environmental exposure or probing of the molecular structure, the induced damage is a major problem that has limited advances in science and technology. We report here the use of an in situ probe that is minimally invasive to detect real-time damage induced in organic materials. Specifically, we use metastable excited helium atoms in the 3S1 state to characterize the damage caused by a low-energy electron beam ∼30 eV on an organic self-assembled monolayer of 11-bromo-1-undecanethiol on a gold substrate. We were able to monitor the damage caused by the electron beam without introducing any additional observed damage by the probing metastable atoms.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
11-Bromo-1-undecanethiol, 99%