Přejít k obsahu
Merck
  • Exploring the Antitumor Mechanisms of Zingiberis Rhizoma Combined with Coptidis Rhizoma Using a Network Pharmacology Approach.
Exploring the Antitumor Mechanisms of Zingiberis Rhizoma Combined with Coptidis Rhizoma Using a Network Pharmacology Approach.
BioMed research international (2021-01-12)
Meng Wang, Youke Qi, Yongning Sun
ANOTACE
Although the combination of Zingiberis rhizoma (ZR) and Coptidis rhizoma (CR) is a classic traditional Chinese medicine-based herbal pair used for its antitumor effect, the material basis and underlying mechanisms are unclear. Here, a network pharmacology approach was used to elucidate the antitumor mechanisms of ZR-CR. To predict the targets of ZR-CR in treating tumors, we constructed protein-protein interactions and hub component-target networks and performed pathway and process enrichment and molecular docking analysis. We used a surface plasmon resonance (SPR) assay to validate the predicted component-target affinities. Hub gene expression and survival analysis in patients with tumors were used to predict the clinical significance. The active components of ZR-CR-shogaol, daucosterol, ginkgetin, berberine, quercetin, chlorogenic acid, and vanillic acid-exhibited antitumor activities via the MAPK, PI3K-AKT, TNF, FOXO, HIF-1, and VEGF signaling pathways. Molecular docking and SPR analyses suggested direct binding of berberine with AKT1 and TP53; quercetin with EGFR and VEGF165; and ginkgetin, isoginkgetin, and daucosterol with VEGF165 with weak affinities. Gene expression levels of the hub targets of ZR-CR were associated with overall survival and disease-free survival in patients with various tumor types. The antitumor components of the ZR-CR herbal pair and the mechanisms underlying their antitumor effects were identified. These antitumor components deserve to be explored further in experimental and clinical studies.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Ginkgetin, phyproof® Reference Substance