Přejít k obsahu
Merck
  • Differentially Expressed Proteins in Intra Synovial Compared to Extra Synovial Flexor Tendon Grafts in a Rabbit Tendon Transplantation Model.
Differentially Expressed Proteins in Intra Synovial Compared to Extra Synovial Flexor Tendon Grafts in a Rabbit Tendon Transplantation Model.
Biomedicines (2020-10-16)
Simon Farnebo, Monica Wiig, Björn Holm, Bijar Ghafouri
ANOTACE
 Uncomplicated healing of grafts for tendon reconstruction remains an unsolved problem in hand surgery. Results are limited by adhesion formation and decreased strength properties, especially within the tight fibro-osseous sheath of the digits. This is especially problematic when an extra synovial tendon graft is used to replace an intra synovial flexor tendon. Compositional differences are likely to play an important role in these processes. The aim of this study was, therefore, to compare protein expression in pair-matched intra synovial tendon grafts with extra synovial tendon grafts, using a rabbit tendon injury model. We hypothesized that there would be significant differences in proteins critical for response to tensile loading and adhesion formation between the two groups. Using mass spectrometry and multivariate statistical data analysis, we found tissue-specific differences in 22 proteins, where 7 explained 93% (R2) of the variation, with a prediction of 81% (Q2). Among the highest discriminating proteins were Galectin, Histone H2A, and Periostin, which were found in a substantially larger amount in the extra synovial tendons compared to the intra synovial tendons. These findings may contribute to improved understanding of the differences in outcome seen after tendon reconstruction using tendon grafts with intra synovial and extra synovial grafts.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Supelco
Cefuroxime, VETRANAL®, analytical standard