Přejít k obsahu
Merck
  • [Pathogenic role leukotriene B4 in lung injury induced by lung-protective mechanical ventilation in rabbits].
[Pathogenic role leukotriene B4 in lung injury induced by lung-protective mechanical ventilation in rabbits].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2020-10-30)
Lingyue Yuan, Jiang Li, Yong Yang, Xin Guo, Xingling Liu, Lisha Li, Xiaoyan Zhu, Rui Liu
ANOTACE
To elucidate the pathogenic role of leukotriene B4 (LTB4) in pulmonary hyper-permeability and inflammation induced by lung-protective mechanical ventilation (LPMV) in rabbits. Thirty-two healthy Japanese white rabbits were randomized into 4 groups for treatment with vehicle or bestatin (a leukotriene A4 hydrolase inhibitor that inhibits LTB4 production) administered intragastrically at the daily dose of 8 mg/kg for 5 days, followed by sham operation (group S and group BS, respectively, in which the rabbits were anesthetized only) or LPMV (group PM and group BPM, respectively, in which the rabbits received ventilation with 50% oxygen at a tidal volume of 8 mL/kg for 5 h). The concentrations of LTB4 and cyclic adenosine monophosphate (cAMP) in the lung tissues were analyzed by ELISA. cAMP content, protein kinase A (PKA) protein expression and the Rap1-GTP protein to total Rap1 protein ratio were determined to assess the activities of cAMP/PKA and Rap1 signaling pathways. The lung injury was evaluated by assessing lung permeability index, lung wet/dry weight ratio, polymorphonuclear leukocyte (PMN) count in bronchoalveolar lavage fluid (BALF), pulmonary myeloperoxidase (MPO) activity and lung histological scores. None of the examined parameters differed significantly between group S and group BS. All the parameters with the exception of lung histological score increased significantly in group PM and group BPM as compared to those in group S (P < 0.05). Compared with those in PM group, the rabbits in group BPM showed significantly reduced LTB4 production in the lungs (P < 0.05), up-regulated cAMP/ PKA and Rap1 signaling pathway activities (P < 0.05), and alleviated lung hyper-permeability and inflammation (P < 0.05). LPMV can induce LTB4 overproduction to down-regulate cAMP/PKA and Rap1 signaling pathways in the lungs of rabbits, which results in lung hyper-permeability and inflammation. Bestatin can inhibit LTB4 production in the lungs to protect against LPMV-induced lung hyper-permeability and inflammation.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
cAMP Enzyme Immunoassay Kit, sufficient for 96 assays