Přejít k obsahu
Merck
SMBA1, a Bax Activator, Induces Cell Cycle Arrest and Apoptosis in Malignant Glioma Cells.
Pharmacology (2019-12-23)
Shuangbo Fan, Qian Xu, Liang Wang, Yulin Wan, Sheng Qiu
ANOTACE
SMBA1 (small-molecule Bax agonists 1), a small molecular activator of Bax, is a potential anti-tumour agent. In the present study, we investigated the biological effects of SMBA1 on glioblastoma (GBM) cells. SMBA1 reduced the viabilities of U87MG, U251 and T98G cells in a time- and dose-dependent manner. Moreover, treatment with SMBA1 induced cell cycle arrest at the G2/M phase transition, accompanied by the downregulation of Cdc25c and cyclin B1 and the upregulation of p21. SMBA1 also induced apoptosis of GBM cells in a dose-dependent manner. Mechanistically, SMBA1 induced apoptosis via the intrinsic pathway. Silencing of Bax or ectopic expression of Bcl-2 significantly inhibited SMBA1-induced apoptosis. Moreover, SMBA1 inhibited the growth of U87MG xenograft tumours in vivo. Overall, SMBA1 shows anti-proliferative effects against GBM cells through activation of the intrinsic apoptosis pathway.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
SMBA1, ≥98% (HPLC)