Přejít k obsahu
Merck
Inflammatory events are involved in acne lesion initiation.
The Journal of investigative dermatology (2003-07-04)
Anthony H T Jeremy, Diana B Holland, Susan G Roberts, Kathryn F Thomson, William J Cunliffe
ANOTACE
The earliest subclinical acne "lesion" is a microcomedone, of which hyperproliferation of the follicular epithelium is a characteristic feature. Inflammatory cells have been observed at the periphery of these "lesions". This study investigated whether inflammatory events occur pre or post hyperproliferative changes. Cellular, vascular, and proliferative markers were examined by immunohistochemical techniques on biopsies of clinically normal follicles from uninvolved skin and early inflamed lesions from acne patients. Control follicles were obtained from non-acne subjects. Follicles from uninvolved skin exhibited no microcomedonal features. Proliferation in the epithelium was comparable to controls and was significantly lower than in inflamed lesions. Numbers of CD3+, CD4+ T cells were elevated in the perifollicular and papillary dermis although levels were not equivalent to those in papules. The number of macrophages was also greatly increased and similar to those in papules. There were no changes in blood vessel numbers or vascular intercellular adhesion molecule 1 expression but E-selectin expression was increased to levels found in papules and vascular adhesion molecule 1 levels were upregulated. Levels of the pro-inflammatory cytokine interleukin-1 were also upregulated perifollicularly. Moreover, aberrant integrin expression was demonstrated in the epidermis around these uninvolved follicles and inflamed lesions whereas the basement membrane was still intact. These results provide novel evidence for vascular endothelial cell activation and involvement of inflammatory responses in the very earliest stages of acne lesion development.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-Integrin αV Antibody, clone P3G8, azide free, clone P3G8, Chemicon®, from mouse