Přejít k obsahu
Merck
  • The Dietary Components Carnosic Acid and Carnosol as Neuroprotective Agents: a Mechanistic View.
The Dietary Components Carnosic Acid and Carnosol as Neuroprotective Agents: a Mechanistic View.
Molecular neurobiology (2016-11-01)
Marcos Roberto de Oliveira
ANOTACE
Carnosic acid (CA) and carnosol are the major diterpenes found in Rosmarinus officinalis (rosemary), a culinary spice. CA and carnosol account for over 90 % of its anti-oxidant activity in rosemary leaves. The diterpenes exert anti-oxidant, anti-inflammatory, and anti-carcinogenic activities, and present neuroprotective effects in both in vitro and in vivo experimental models. In some cases, CA exerted protective effects upon neuronal cells more intensely than resveratrol or sulforaphane. Therefore, CA and carnosol demonstrate a potential pharmacological role for rosemary diterpenes in ameliorating mammalian brain redox status, among other parameters, as for instance the modulation of neuroinflammation. The aim of this review is to discuss the biological effects of CA and carnosol on neuronal and glial cells with focus on the mechanism of action of such diterpenes.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Carnosol