Přejít k obsahu
Merck
  • Fermented and nonfermented liquid feed to growing pigs: effect on aspects of gastrointestinal ecology and growth performance.
Fermented and nonfermented liquid feed to growing pigs: effect on aspects of gastrointestinal ecology and growth performance.
Journal of animal science (2003-08-21)
N Canibe, B B Jensen
ANOTACE
The effect of feeding dry feed (DF), nonfermented liquid feed (NFLF), and fermented liquid feed (FLF) to growing pigs on aspects of gastrointestinal ecology and on performance was investigated. Nonfermented liquid feed was prepared by mixing feed and water at a ratio of 1:2.5 immediately before feeding. Fermented liquid feed was prepared by mixing feed and water in the same ratio as NFLF, and stored in a tank at 20 degrees C for 4 d, after which half the volume was removed twice daily at each feeding and replaced with the same volume of feed and water mixture. A total of 60 pigs (initial BW of 30.7 kg) from 20 litters was used. Twenty pigs, housed individually, were allotted to each of the diets and fed restrictively. Five pigs from each diet were sacrificed at an average BW of 112 kg and digesta from the gastrointestinal tract (GI-tract) was obtained to examine variables describing some aspects of the gastrointestinal ecology. Fermented liquid feed contained high levels of lactic acid bacteria (9.4 log cfu/g) and lactic acid (approximately 169 mmol/kg), low levels of enterobacteria (<3.2 log cfu/g), and had a low pH (4.4). Nonfermented liquid feed contained 7.2 log cfu/g of lactic acid bacteria, and 6.2 log cfu/g of enterobacteria, which indicated that fermentation had started in the feed. The pigs fed FLF had the lowest levels of enterobacteria along the GI-tract (<3.2 to 5.0 log cfu/g), and those fed NFLF the highest levels (5.7 to 6.6 log cfu/g; P < or = 0.02). Fermented liquid feed caused a decrease in gastric pH from 4.4 and 4.6 for DF and NLF, to 4.0 (P = 0.003), and increased numerically the gastric concentration of lactic acid (P = 0.17) from 50 to 60 mmol/kg in the DF and NFLF treatments to 113 mmol/kg in the FLF treatment. The animals fed NFLF showed the highest weight gain (995 g/d) and feed intake (2.14 kg/d), and those fed FLF the lowest values (weight gain, 931 g/d; feed intake, 1.96 kg/d; P = 0.003 for weight gain, and P < 0.001 for feed intake). The results from the present study indicate that feeding FLF as prepared here may be a valid feeding strategy to decrease the levels of enterobacteria in the GI-tract of growing pigs, whereas feeding liquid feed that has started to ferment (high levels of enterobacteria and high pH as with NFLF) increases the presence of these undesirable bacteria. Nonetheless, higher daily feed intake and body weight gain are obtained when feeding NFLF compared with feeding FLF or DF.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-GRK 2/3 (βARK 1/2) Antibody, clone C5/1.1, clone C5/1.1, Upstate®, from mouse