Přejít k obsahu
Merck
  • Geographic location determines beta-cell autoimmunity among adult Ghanaians: Findings from the RODAM study.
Geographic location determines beta-cell autoimmunity among adult Ghanaians: Findings from the RODAM study.
Immunity, inflammation and disease (2020-05-08)
Christiane S Hampe, Diomira Sahabandu, Vivien Kaiser, Tanja Telieps, Liam Smeeth, Charles Agyemang, Joachim Spranger, Matthias B Schulze, Frank P Mockenhaupt, Ina Danquah, Olov Rolandsson
ANOTACE
Beta-cell autoantibodies are established markers of autoimmunity, which we compared between Ghanaian adults with or without diabetes, living in rural and urban Ghana and in three European cities. In the multicenter cross-sectional Research on Obesity and Diabetes among African Migrants (RODAM) study (N = 5898), we quantified autoantibodies against glutamic acid decarboxylase (GAD65Ab) by radioligand binding assay (RBA) and established cut-offs for positivity by displacement analysis. In a subsample, we performed RBA for zinc transporter-8 autoantibodies (ZnT8Ab). Associations of environmental, sociodemographic, and clinical factors with GAD65Ab were calculated. In this study population (age: 46.1 ± 11.9 years; female: 62%; Ghana-rural: 1111; Ghana-urban: 1455; Europe: 3332), 9.2% had diabetes with adult-onset. GAD65Ab concentrations were the highest in Ghana-rural (32.4; 10.8-71.3 U/mL), followed by Ghana-urban (26.0; 12.3-49.1 U/mL) and Europe (11.9; 3.0-22.8 U/mL) with no differences between European cities. These distributions were similar for ZnT8Ab. Current fever, history of fever, and higher concentrations of liver enzymes marginally explained site-specific GAD65Ab concentrations. GAD65Ab positivity was as frequent in diabetes as in nondiabetes (5.4% vs 6.1%; P = .25). This was also true for ZnT8Ab positivity. Geographic location determines the occurrence of GAD65Ab and ZnT8Ab more than the diabetes status. Beta-cell autoimmunity may not be feasible to differentiate diabetes subgroups in this population.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Mouse IgG2a ELISA Kit