Přejít k obsahu
Merck
  • iTRAQ-based proteomics implies inflammasome pathway activation in the prefrontal cortex of CSDS mice may influence resilience and susceptibility.
iTRAQ-based proteomics implies inflammasome pathway activation in the prefrontal cortex of CSDS mice may influence resilience and susceptibility.
Life sciences (2020-09-30)
Tianlan Lan, Mengge Bai, Xiangyu Chen, Yue Wang, Yan Li, Yu Tian, Yong He, Zhonghao Wu, Heming Yu, Zhi Chen, Chong Chen, Yaping Yu, Ke Cheng, Peng Xie
ANOTACE
Major depressive disorder, as a destructive mental health disorder, is a major contributor to disability and death. Numerous studies have illustrated that activation of inflammation and fluctuating immune reactions play a crucial role in the physiopathology of depression. The effectiveness of antidepressants is affected by the intensity of the inflammatory response. Thus, we aim to reveal the correlation of inflammatory factors and depression. Isobaric tags for relative and absolute quantitation (iTRAQ™)-based proteomics was applied to verify the quantitation of target proteins in the PFC of chronic social defeat stress (CSDS) model mice. Ingenuity pathway analysis (IPA) was performed to explore related pathways, and the involvement of molecules was validated by western blotting and real time-quantitative polymerase chain reaction (RT-qPCR). According to the IPA results, CSDS-susceptible mice and CSDS-resilient mice both exhibited alterations of the inflammasome pathway in the PFC. Compared with control mice, susceptible mice subjected to CSDS showed an increased ATP-activated purinergic receptor P2X7 (also known as P2RX7) protein level. Nevertheless, the expression levels of cysteinyl aspartate-specific protease 1 (Caspase 1) and apoptosis-associated speck-like protein containing a CARD (ASC) were reduced in CSDS mice, and downregulation of interleukin-1β (IL-1β) was found in susceptible mice. Moreover, no significant difference was found in nuclear factor-κB levels among the three groups. CSDS administration leads to dysfunctions of key molecules in the inflammasome pathway, promoting depressive-like behaviors in mice.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Supelco
Empore Extraction Disk Cartridge, matrix active group C18-SD, volume 3 mL, pk of 50